Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Mercury(II) Amidochloride

Catalog No. 10005500GR Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-005-500GR 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 10-005-500GR Supplier Sigma Aldrich Supplier No. 10005500GR
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Mercury(II) Amidochloride

Specifications

CAS 1067-87-4
Boiling Point 46°C (0.35 mmHg, lit.)
Molecular Formula C7H15O3P
Linear Formula CH2=CHCH2P(O)(OC2H5)2
MDL Number MFCD00015134
Quantity 5 g
Synonym Amidomercury(II) chloride
Molecular Weight (g/mol) 178.17 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.