Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) Bromide, ≥99.0% (precipitation titration), Puriss. p.a., ACS Reagent, Solstice

Catalog No. 60047654 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-654 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-654 Supplier Solstice Supplier No. 8335350G
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent,≥99.0% (precipitation titration)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-47-1
Molecular Formula Br2Hg
Molecular Weight (g/mol) 360.4
MDL Number MFCD00011040
InChI Key NGYIMTKLQULBOO-UHFFFAOYSA-L
Synonym mercuric bromide, mercury dibromide, mercury ii bromide, hgbr2, mercury bromide hgbr2, mercuric dibromide, mercury ii bromide 1:2, hsdb 829, hg2, mercury 2 bromide
PubChem CID 24612
ChEBI CHEBI:49639
IUPAC Name dibromomercury
SMILES Br[Hg]Br

Specifications

Boiling Point 322°C
Melting Point 237°C
Color Violet to White
pH 4.5 (at 5.00g/L)
Physical Form Solid
Quantity 50 g
Linear Formula HgBr2
UN Number UN1634
Formula Weight 360.4g/mol
Percent Purity ≥99.0% (precipitation titration)
Odor Odorless
Packaging Glass bottle
Vapor Pressure 1mmHg (136.5°C)
Density 6.1g/cu.cm at 25°C
Chemical Name or Material Mercury(II) bromide
Show More Show Less
Hazard Statement Missing Phrase - N15.00950417-P260-P262-P280-P302 + P352 + P310-P304 + P340 + P310
DOTInformation 6.1/PGII
RTECSNumber 232-169-3
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.