Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) bromide, 99+%

Catalog No. AC190495000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC190495000 500 g Glass bottle
AC190491000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. AC190495000 Supplier Thermo Scientific Chemicals Supplier No. 190495000
  • $372.00
    $316.65 / Each of 1

    Save $55.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

CAS: 7789-47-1 | Br2Hg | 360.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-47-1
Molecular Formula Br2Hg
Molecular Weight (g/mol) 360.4
MDL Number MFCD00011040
InChI Key NGYIMTKLQULBOO-UHFFFAOYSA-L
Synonym mercuric bromide, mercury dibromide, mercury ii bromide, hgbr2, mercury bromide hgbr2, mercuric dibromide, mercury ii bromide 1:2, hsdb 829, hg2, mercury 2 bromide
PubChem CID 24612
ChEBI CHEBI:49639
IUPAC Name dibromomercury
SMILES Br[Hg]Br

Specifications

Boiling Point 322.0°C
Melting Point 236.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 500 g
Assay Percent Range 99% min
Linear Formula HgBr2
Fieser 05,324
Merck Index 15, 5942
Solubility Information Solubility in water: 0.6g/100mL (25 c). Other solubilities: freely soluble in hcl and hbr, soluble in alcohol and ether, slightly soluble in chloroform
Formula Weight 360.4
Percent Purity ≥99%
Packaging Glass bottle
Density 6.1
Chemical Name or Material Mercury(II) bromide, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal in contact with skin.
Fatal if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Fatal if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 232-169-3
RTECSNumber OV7415000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.