Learn More
Strem, An Ascensus Company CAS# 7789-47-1. 50g. Mercury(II) bromide (ACS). MFCD00011040
SDP
Catalog No. 5090116399
Encompass
CAS# 7789-47-1. 50g. Mercury(II) bromide (ACS). MFCD00011040. Molecular Weight: 360.44. Molecular Formula: HgBr2. Color/form: white pwdr. Strem# 93-8003. http://www.strem.com/catalog/v/93-8003/
Catalog No. 50-901-16399 Supplier Strem, An Ascensus Company Supplier No. 93800350G
CAS# 7789-47-1. 50g. Mercury(II) bromide (ACS). MFCD00011040. Molecular Weight: 360.44. Molecular Formula: HgBr2. Color/form: white pwdr. Strem# 93-8003. http://www.strem.com/catalog/v/93-8003/