Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) bromide, ACS

Catalog No. AA4481622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4481622 100 g
AA4481614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4481622 Supplier Thermo Scientific Chemicals Supplier No. 04481622
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7789-47-1 | Br2Hg | 360.4 g/mol

Mercury(II) bromide is used as a reagent in the Koenigs-Knorr reaction, which forms glycoside linkages on carbohydrates. It is also used to test for the presence of arsenic, as recommended by the Pharmacopoeia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-47-1
Molecular Formula Br2Hg
Molecular Weight (g/mol) 360.4
MDL Number MFCD00011040
InChI Key NGYIMTKLQULBOO-UHFFFAOYSA-L
Synonym mercuric bromide, mercury dibromide, mercury ii bromide, hgbr2, mercury bromide hgbr2, mercuric dibromide, mercury ii bromide 1:2, hsdb 829, hg2, mercury 2 bromide
PubChem CID 24612
ChEBI CHEBI:49639
IUPAC Name dibromomercury
SMILES Br[Hg]Br

Specifications

Boiling Point 322°C to 325°C (sublimation)
Melting Point 236°C
Color White
Physical Form Powder
Quantity 100 g
Odor Odorless
Sensitivity Light sensitive
Vapor Pressure 1mm Hg at 137°C
Density 6.05
Solubility Information Soluble in hot alcohol,methanol,HCl,HBr
Formula Weight 360.4
Grade ACS
Chemical Name or Material Mercury(II) bromide, For ACS analysis
Show More Show Less
Hazard Category H300+H310+H330-H373
Precautionary Statement P260-P262-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 232-169-3
RTECSNumber OV7415000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.