Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) chloride, 98+%

Catalog No. AAA174720B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA174720B 1000 g
AAA1747218 50 g
AAA1747230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA174720B Supplier Thermo Scientific Chemicals Supplier No. A174720B
Add to Cart

CAS: 7487-94-7 | Cl2Hg | 271.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
The main application of mercuric chloride is as a catalyst for the conversion of acetylene to vinyl chloride, the precursor to polyvinylchloride. It is also employed in photography, disinfectants, wood preservatives

Solubility
Soluble in diethyl ether. Insoluble in water.

Notes
Store at room temperature. Reactive with oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-94-7
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
PubChem CID 24085
ChEBI CHEBI:31823
IUPAC Name dichloromercury
SMILES [Cl-].[Cl-].[Hg++]

Specifications

Boiling Point 302°C
Melting Point 280°C to 282°C
Quantity 1000 g
UN Number UN1624
Merck Index 14,5876
Solubility Information Soluble in diethyl ether. Insoluble in water.
Formula Weight 271.5
Percent Purity ≥98%
Odor Odorless
Refractive Index 1.859
Density 5.4
Chemical Name or Material Mercury(II) chloride
Show More Show Less
Hazard Category H300-H310-H314-H335-H341-H361f-H372
Hazard Statement GHS H Statement
H300-H372-H341-H361-H314
Fatal if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Precautionary Statement P201-P202-P260-P262-P264b-P270-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURIC CHLORIDE
EINECSNumber 231-299-8
RTECSNumber OV9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.