Learn More
Mercury(II) chloride, 98+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
The main application of mercuric chloride is as a catalyst for the conversion of acetylene to vinyl chloride, the precursor to polyvinylchloride. It is also employed in photography, disinfectants, wood preservatives
Solubility
Soluble in diethyl ether. Insoluble in water.
Notes
Store at room temperature. Reactive with oxidizing agents and strong bases.
Specifications
Specifications
| Boiling Point | 302°C |
| Melting Point | 280°C to 282°C |
| Quantity | 50 g |
| UN Number | UN1624 |
| Merck Index | 14,5876 |
| Solubility Information | Soluble in diethyl ether. Insoluble in water. |
| Formula Weight | 271.5 |
| Percent Purity | ≥98% |
| Odor | Odorless |
| Refractive Index | 1.859 |
| Show More |
Safety and Handling
| Hazard Category | H300-H310-H314-H335-H341-H361f-H372 |
| Hazard Statement | GHS H Statement H300-H372-H341-H361-H314 Fatal if swallowed. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Suspected of damaging fertility or the unborn child. Causes severe skin burns and eye damage. |
| Precautionary Statement | P201-P202-P260-P262-P264b-P270-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURIC CHLORIDE |
| EINECSNumber | 231-299-8 |
| RTECSNumber | OV9100000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.