Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(Ii) Chloride, ACS Reagent, Solstice

Catalog No. 6003245 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-032-45 500 g
60-032-46 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-032-45 Supplier Solstice Supplier No. 215465500G
Add to Cart
Add to Cart

ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-94-7
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
PubChem CID 24085
ChEBI CHEBI:31823
IUPAC Name dichloromercury
SMILES [Cl-].[Cl-].[Hg++]

Specifications

Physical Form Powder
Quantity 500 g
Linear Formula HgCl2
UN Number 1624
Solubility Information Soluble in diethyl ether, acetic acid, ethyl acetate, glycerol, alcohol, acetone (salt form), water
Formula Weight 271.5g/mol
Percent Purity ≥99.5%
Grade ACS Reagent
Vapor Pressure 1.3mmHg (236°C)
Chemical Name or Material Mercury(Ii) Chloride
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H300 + H310-H314-H341-H361f-H372-H410
EINECSNumber 231-299-8
RTECSNumber OV9100000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.