Learn More
Strem, An Ascensus Company CAS# 592-04-1. 50g. Mercury(II) cyanide, 99%. MFCD00011037
SDP
Catalog No. 5090116434
Encompass
CAS# 592-04-1. 50g. Mercury(II) cyanide, 99%. MFCD00011037. Molecular Weight: 252.65. Molecular Formula: Hg(CN)2. Color/form: white pwdr. Strem# 93-8041. http://www.strem.com/catalog/v/93-8041/
Catalog No. 50-901-16434 Supplier Strem, An Ascensus Company Supplier No. 93804150G
CAS# 592-04-1. 50g. Mercury(II) cyanide, 99%. MFCD00011037. Molecular Weight: 252.65. Molecular Formula: Hg(CN)2. Color/form: white pwdr. Strem# 93-8041. http://www.strem.com/catalog/v/93-8041/