Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) iodide, ACS, 99.0% min (Assay-dried basis)

Catalog No. p-4407640
Click to view available options
Quantity:
100 g
500 g

For Use As An Analytical Reagent For the Preparation of Nessler'S Reagent. | CAS: 7774-29-0 | HgI2 | 454.401 g/mol

As an analytical reagent for the preparation of Nessler's Reagent. It is also used as an oxidizing agent for the synthesis of quinoxaline. Polycrystalline films of mercury (II) iodide, also play an important role in X-ray imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7774-29-0
Molecular Formula HgI2
Molecular Weight (g/mol) 454.401
MDL Number MFCD00011044
InChI Key YFDLHELOZYVNJE-UHFFFAOYSA-L
Synonym mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum
PubChem CID 24485
ChEBI CHEBI:49659
IUPAC Name diiodomercury
SMILES I[Hg]I

Specifications

Boiling Point 350°C (sublimation)
Melting Point 259°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.0% min.
Solubility Information Insoluble in water. Soluble in boiling alcohol,ether,acetone,ethyl acetate,carbon disulfide,alkali iodides.
Formula Weight 454.4
Grade ACS
Odor Odorless
Sensitivity Light sensitive
Appearance Red powder or beads
Density 6.28 g/cm3 at 20°C
Chemical Name or Material Mercury(II) iodide, Assay-dried basis
Show More Show Less
Hazard Category H300+H310+H330-H373
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 231-873-8
RTECSNumber OW5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.