Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(Ii) Iodide, Solstice

Catalog No. 6003250 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-032-50 25 g
60-032-49 100 g
60-032-51 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-032-50 Supplier Solstice Supplier No. 22109025G
Add to Cart
Add to Cart

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7774-29-0
Molecular Formula HgI2
Molecular Weight (g/mol) 454.401
MDL Number MFCD00011044
InChI Key YFDLHELOZYVNJE-UHFFFAOYSA-L
Synonym mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum
PubChem CID 24485
ChEBI CHEBI:49659
IUPAC Name diiodomercury
SMILES I[Hg]I

Specifications

Quantity 25 g
Linear Formula HgI2
UN Number 1638
Solubility Information Soluble in Potassium Iodide solution
Formula Weight 454.4g/mol
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Mercury(Ii) Iodide
Hazard Statement P260-P262-P280-P302 + P352 + P310-P304 + P340 + P310
Precautionary Statement H300 + H310 + H330-H373-H410
EINECSNumber 231-873-8
RTECSNumber OW5250000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.