Learn More
Strem, An Ascensus Company CAS# 7783-34-8. 50g. Mercury(II) nitrate monohydrate, 98+% (ACS). MFCD00149736
SDP
Catalog No. 5090116407
Encompass
CAS# 7783-34-8. 50g. Mercury(II) nitrate monohydrate, 98+% (ACS). MFCD00149736. Molecular Weight: 324.60 (342.62). Molecular Formula: Hg(NO3)2∙H2O. Color/form: white xtl. Strem# 93-8009. http://www.strem.com/catalog/v/93-8009/
Catalog No. 50-901-16407 Supplier Strem, An Ascensus Company Supplier No. 93800950G
CAS# 7783-34-8. 50g. Mercury(II) nitrate monohydrate, 98+% (ACS). MFCD00149736. Molecular Weight: 324.60 (342.62). Molecular Formula: Hg(NO3)2∙H2O. Color/form: white xtl. Strem# 93-8009. http://www.strem.com/catalog/v/93-8009/