Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(Ii) Oxide Red, ACS Reagent, ≥99.0%, Solstice

Catalog No. 60045225 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-225 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-225 Supplier Solstice Supplier No. 213357100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21908-53-2
Molecular Formula HgO
Molecular Weight (g/mol) 216.59
MDL Number MFCD00011045
InChI Key UKWHYYKOEPRTIC-UHFFFAOYSA-N
Synonym mercuric oxide, mercury ii oxide, mercury oxide, mercuric oxide, red, red mercuric oxide, mercuric oxide, yellow, santar, red precipitate, mercury monoxide
IUPAC Name oxomercury
SMILES O=[Hg]

Specifications

Melting Point >400°C
Physical Form Powder
Quantity 100 g
Linear Formula HgO
UN Number UN1641
Formula Weight 216.59g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Glass Bottle
Appearance Powder
Chemical Name or Material Mercury(Ii) Oxide Red
Show More Show Less
Hazard Statement Missing Phrase - N15.00950417-P260-P262-P280-P302 + P352 + P310-P304 + P340 + P310
Precautionary Statement H300 + H310 + H330-H373-H410
DOTInformation 6.1/PGII
EINECSNumber 244-654-7
RTECSNumber OW8750000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.