Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) potassium iodide, Thermo Scientific Chemicals

Catalog No. AA3930730
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3930730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3930730 Supplier Thermo Scientific Chemicals Supplier No. 03930730
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7783-33-7 | Molecular Formula: H4HgI4K2 | Molecular Weight: 790.44 g/mol

Mercury(II) potassium iodide is used as a pharmaceutical intermediate. It is also used as reagents, chemical research and organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-33-7
Molecular Formula H4HgI4K2
Molecular Weight (g/mol) 790.44
MDL Number MFCD00049656
InChI Key ZCPVFAXYZYZMBC-UHFFFAOYSA-N
Synonym nessler's reagent, nessler reagent, mayer's reagent, channing's solution, caswell no. 695, potassium iodomercurate, mercury ii potassium iodide, mercury potassium iodide, potassium mercuric iodide, mercuric potassium iodide
PubChem CID 24542
ChEBI CHEBI:51568
IUPAC Name dipotassium;tetraiodomercury(2-)
SMILES [K+].[K+].I.I.I.I.[Hg--]

Specifications

Color Yellow
Physical Form Crystalline Aggregates
Quantity 250 g
Linear Formula K2HgI4
UN Number UN2025
Merck Index 14,7688
Solubility Information Soluble in water.
Formula Weight 786.4
Sensitivity Hygroscopic
Density 4.3 g/mL
Chemical Name or Material Mercury(II) potassium iodide
Show More Show Less
Hazard Category H300+H310+H330-H373
Hazard Statement GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
EINECSNumber 231-990-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.