Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) selenide, 99.9% (metals basis)

Catalog No. AA2287804
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2287804 2 g
AA2287809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2287804 Supplier Thermo Scientific Chemicals Supplier No. 02287804
Add to Cart

CAS: 20601-83-6 | HgSe | 279.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20601-83-6
Molecular Formula HgSe
Molecular Weight (g/mol) 279.56
MDL Number MFCD00049743
InChI Key YQMLDSWXEQOSPP-UHFFFAOYSA-N
Synonym mercury selenide, mercury ii selenide, mercury-selenium complex, mercury monoselenide, mercury 2+ selenide, mercury ii selenide trace metals basis 1g
PubChem CID 88609
IUPAC Name selanylidenemercury
SMILES [Se]=[Hg]

Specifications

Melting Point (sublimation)
Color Gray
Physical Form Powder
Quantity 2 g
UN Number UN2025
Formula Weight 279.55
Percent Purity 99.9%
Odor Odorless
Density 8.21 g/mL
Assay (metals basis)
Chemical Name or Material Mercury(II) selenide
Show More Show Less
Hazard Category H301-H330-H360Df-H373
Hazard Statement GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
EINECSNumber 243-910-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.