Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) sulfate, 98+%

Catalog No. AAA1633018
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1633018 50 g
AAA1633030 250 g
AAA163300B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1633018 Supplier Thermo Scientific Chemicals Supplier No. A1633018
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7783-35-9 | HgO4S | 296.65 g/mol

It is used in medicine as a catalyst and in extracting gold and silver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine as a catalyst and in extracting gold and silver.

Solubility
It is soluble in hot H2SO4, NaCl solution. insoluble in alcohol, acetone, ammonia

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Light sensitive. Moisture sensitive. Air sensitive. Keep away from Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
PubChem CID 24544
SMILES [Hg++].[O-]S([O-])(=O)=O

Specifications

Melting Point (decomposition)
Quantity 50 g
Linear Formula HgSO4
UN Number UN1645
Merck Index 14,5887
Solubility Information It is soluble in hot H2SO4,NaCl solution. insoluble in alcohol,acetone,ammonia
IUPAC Name mercury(2+);sulfate
Formula Weight 296.65
Percent Purity ≥98%
Sensitivity Moisture sensitive
Density 6.47
Chemical Name or Material Mercury(II) sulfate
Show More Show Less
Hazard Category H300+H310+H330-H373
Hazard Statement GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY SULFATES
EINECSNumber 231-992-5
RTECSNumber OX0500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.