Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) sulfate, 99+%

Catalog No. AC190500025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC190500025 2.5 kg Plastic bottle
AC190501000 100 g Plastic bottle
AC190505000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC190500025 Supplier Thermo Scientific Chemicals Supplier No. 190500025

CAS: 7783-35-9 | HgO4S | 296.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
PubChem CID 24544
SMILES [Hg++].[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 99% min
Linear Formula HgSO4
Fieser 01,658; 15,2
Merck Index 15,5955
Solubility Information Solubility in water: reacts. Other solubilities: soluble in hcl, hot dil. h2so4 and sol. of nacl
IUPAC Name mercury(2+) sulfate
Formula Weight 296.64
Percent Purity ≥99%
Packaging Plastic bottle
Specific Gravity 6.47
Density 6.4700g/mL
Chemical Name or Material Mercury(II) sulfate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Fatal in contact with skin.
Fatal if swallowed.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash w
EINECSNumber 231-992-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.