Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) sulfate, ACS, 98.0% min

Catalog No. AA3628622
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3628622 100 g
AA3628614 25 g
AA3628636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3628622 Supplier Thermo Scientific Chemicals Supplier No. 03628622
  • $176.80
    $150.65 / Each of 1

    Save $26.15

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

For Use As a Primary Battery Electrolyte, Catalyst in Conversion of Acetylene To Acetaldehyde | CAS: 7783-35-9 | HgO4S | 296.65 g/mol

Mercury(II) sulfate is used for gold and silver extraction, and to make other mercury compounds. Used as catalyst in the conversion of acetylene to acetaldehyde and used as a electrolyte in primary battery. It is commonly used as a catalyst in oxymercuration-demercuration, a type of electrophilic addition reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
PubChem CID 24544
IUPAC Name mercury(2+);sulfate
SMILES [Hg++].[O-]S([O-])(=O)=O

Specifications

Melting Point (decomposition)
Color White to Yellow
Physical Form Powder
Quantity 100 g
Assay Percent Range 98% min.
Solubility Information Soluble in hot sulfuric acid and sodium chloride solution. Insoluble in alcohol,acetone and ammonia.
Formula Weight 296.65
Grade ACS
Sensitivity Hygroscopic
Appearance White to pale yellow powder
Density 6.470 g/cm3 at 20°C
Chemical Name or Material Mercury(II) sulfate
Show More Show Less
Hazard Category H300+H310+H330-H373
Hazard Statement Keep away from food, drink and animal feeding stuffs. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P501c
DOTInformation DOT Class: 6.1; Packing Group: II
EINECSNumber 231-992-5
RTECSNumber OX0500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.