Learn More
Mercury(II) sulfate, ACS, 98.0% min
Description
Mercury(II) sulfate is used for gold and silver extraction, and to make other mercury compounds. Used as catalyst in the conversion of acetylene to acetaldehyde and used as a electrolyte in primary battery. It is commonly used as a catalyst in oxymercuration-demercuration, a type of electrophilic addition reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Color | White to Yellow |
| Physical Form | Powder |
| Quantity | 500 g |
| Sensitivity | Hygroscopic |
| Appearance | White to pale yellow powder |
| Assay Percent Range | 98% min. |
| Density | 6.47 |
| Solubility Information | Soluble in hot sulfuric acid and sodium chloride solution. Insoluble in alcohol,acetone and ammonia. |
| Formula Weight | 296.65 |
| Show More |
Safety and Handling
| Hazard Category | H300+H310+H330-H373 |
| Hazard Statement | Keep away from food, drink and animal feeding stuffs. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets. |
| Precautionary Statement | P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P501c |
| DOTInformation | DOT Class: 6.1; Packing Group: II |
| EINECSNumber | 231-992-5 |
| RTECSNumber | OX0500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.