Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, suitable as ACS-grade LC reagent,, ≥99.9%, MilliporeSigma™ Supelco™

Catalog No. 111017920 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7920 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7920 Supplier MilliporeSigma Supelco Supplier No. 439193200L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Food and Beverages
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00004595
Synonym Methyl alcohol

Specifications

Melting Point -98°C
CAS 67-56-1
Density 0.791 g/mL (at 25°C)
Quantity 200 L
Ignition Point 725°F
Linear Formula CH3 OH
Vapor Pressure 410 mmHg (50°C)
Beilstein 1098229
Boiling Point 64.7°C
Formula Weight 32.04
Percent Purity ≥99.9%
Grade HPLC Gradient
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.