Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) sulfate, Solstice

Catalog No. 6003258 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-032-58 3 kg
60-032-55 100 g
60-032-57 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-032-58 Supplier Solstice Supplier No. 833843KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent,≥99.0% (precipitation titration)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-35-9
Molecular Formula HgO4S
Molecular Weight (g/mol) 296.65
MDL Number MFCD00011047
InChI Key DOBUSJIVSSJEDA-UHFFFAOYSA-L
Synonym mercury ii sulfate, mercuric sulfate, mercury sulphate, mercury bisulfate, mercury disulfate, mercury persulfate, mercuric sulphate, mercury sulfate hgso4, unii-j4l3ppg58i, mercuric bisulphate
PubChem CID 24544
IUPAC Name mercury(2+);sulfate
SMILES [Hg++].[O-]S([O-])(=O)=O

Specifications

Quantity 3 kg
Linear Formula HgSO4
UN Number UN1645
Formula Weight 296.65g/mol
Percent Purity ≥99% (precipitation titration)
Chemical Name or Material Mercury(II) sulfate
Hazard Statement P260-P262-P280-P302 + P352 + P310-P304 + P340 + P310
RTECSNumber OX0500000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.