Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) telluride, 99.999% (metals basis)

Catalog No. AA3666609
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3666609 10 g
AA3666604 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3666609 Supplier Thermo Scientific Chemicals Supplier No. 03666609
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 12068-90-5 | HgTe | 328.19 g/mol

Mercury cadmium telluride (MCT)is the mostim- portantmaterial currently in use for the manufacture of high performance infrared (IR) detectors. Mercury telluride and mercury telluride-cadmium telluride are semiconductors which have been investigated as intrinsic infrared detecting materials. Complete coverage of the low-loss fiber windows by mercury telluride quantum dot are used in colloidal nanocrystals for telecommunications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12068-90-5
Molecular Formula HgTe
Molecular Weight (g/mol) 328.19
MDL Number MFCD00016133
InChI Key VCEXCCILEWFFBG-UHFFFAOYSA-N
Synonym mercury telluride, mercury ii telluride, mercuric telluride, quecksilbertellurid, mercurous telluride, mercury ii telluride trace metals basis 5g
PubChem CID 82914
IUPAC Name tellanylidenemercury
SMILES [Te]=[Hg]

Specifications

Melting Point 673°C
Color Black to Gray
Physical Form Fused Lump
Quantity 10 g
Odor Odorless
Density 8.63 g/mL
UN Number UN2025
Solubility Information Insoluble in water.
Formula Weight 328.19
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Mercury(II) telluride
Show More Show Less
Hazard Category H300+H310+H330-H373
Hazard Statement GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
EINECSNumber 235-108-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.