Learn More
Mercury(II) telluride, 99% (metals basis), Thermo Scientific Chemicals
Description
Mercury telluride in the photoelectromagnetic-Nernst configuration exhibit spectral and frequency responses indicative of thermal rather than photon behavior.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 673°C |
| Color | Black to Gray |
| Physical Form | Powder |
| Quantity | 5 g |
| Odor | Odorless |
| Density | 8.63 g/mL |
| UN Number | UN2025 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 328.19 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Hazard Category | H300+H310+H330-H373 |
| Hazard Statement | GHS H Statement H300-H310-H330-H373 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S. |
| EINECSNumber | 235-108-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.