Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) thiocyanate, 99+%

Catalog No. AC196892500
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC196892500 250 g Glass bottle
AC196890500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC196892500 Supplier Thermo Scientific Chemicals Supplier No. 196892500
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 592-85-8 | C4HgK2N4S4 | 511.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-85-8
Molecular Formula C4HgK2N4S4
Molecular Weight (g/mol) 511.10
MDL Number MFCD00011039
InChI Key HWXOCCNUUYRBKP-UHFFFAOYSA-J
IUPAC Name mercury(2+) dipotassium tetrakis(cyanosulfanide)
SMILES [K+].[K+].[Hg++].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N

Specifications

Melting Point 165.0°C
Color White
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula Hg(SCN)2
Flash Point 120°C
Fieser 09,293
Merck Index 15, 5958
Solubility Information Solubility in water: 0.7g/L (25°C). Other solubilities: soluble in diluted hcl
Formula Weight 316.75
Percent Purity ≥99%
Packaging Glass bottle
Chemical Name or Material Mercury(II) thiocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Fatal if swallowed.
Fatal in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at r
EINECSNumber 209-773-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.