Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury(II) trifluoroacetate, 98+%

Catalog No. AA8813206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8813206 5 g
AA8813214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8813206 Supplier Thermo Scientific Chemicals Supplier No. 08813206
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13257-51-7 | C4F6HgO4 | 426.622 g/mol

Mercury(II) trifluoroacetate promotes selective cyclization of methylthio-substituted divinyl ketones to 5-fluorocyclopentenone derivatives. It is a raw material of fine chemicals and pharmaceutical intermediates. So it can be used in the pharmaceutical synthesis. Both linear and cross-linked copolymers of N-vinylcarbazole and methylacrylate can be prepared and mercuriated by using mercury trifluoroacetate in THF or dichloromethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13257-51-7
Molecular Formula C4F6HgO4
Molecular Weight (g/mol) 426.622
MDL Number MFCD00013201
InChI Key WMHOESUUCMEQMS-UHFFFAOYSA-L
Synonym mercury ii trifluoroacetate, mercury bis trifluoroacetate, mercury 2+ ditrifluoroacetate, mercury 2+ ; 2,2,2-trifluoroacetate, bis trifluoroacetato mercury, hg ococf3 2, hg o2ccf3 2, mercury bis-trifluoroacetate, mercury ii bis trifluoroacetate
PubChem CID 2723924
IUPAC Name mercury(2+);2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Hg+2]

Specifications

Melting Point 167°C to 169°C
Color White
Linear Formula Hg(OOCCF3)2
Quantity 5 g
UN Number UN2025
Beilstein 3633860
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 426.62
Percent Purity ≥98%
Physical Form Crystalline
Chemical Name or Material Mercury (II) trifluoroacetate
Show More Show Less
Hazard Category H300+H310+H330-H373
Hazard Statement GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
EINECSNumber 236-250-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.