Learn More
Perkin Elmer US LLC Mercury Lamp for PerkinElmer Flow Injection Mercury System (FIMS)
Catalog No. 509042542
Encompass_Preferred
This is a replacement Mercury (Hg) Source Lamp for the PerkinElmer Flow Injection Mercury System (FIMS) series of instruments. Safety requires that the lamp extraction tool, B3000243, be used during its replacement.
Catalog No. 50-904-2542 Supplier Perkin Elmer US LLC Supplier No. B0510487
This is a replacement Mercury (Hg) Source Lamp for the PerkinElmer Flow Injection Mercury System (FIMS) series of instruments. Safety requires that the lamp extraction tool, B3000243, be used during its replacement.