Learn More
MSC Kwazar Mercury Pro Bottle, Dual Action, Viton Seals, 1.0 L
Catalog No. 502866098
Encompass
The Kwazar Mercury Pro+ Spray Bottle features a robust, ergonomically designed construction for professional applications. It is equipped with a unique dual-action trigger that provides a constant, fine mist with reduced hand fatigue, suitable for various chemical dispensing tasks.
  • Unique double-action trigger provides a constant, fine spray
  • Viton seals resistant to most common chemicals and detergents
  • Level indicator embossed on container marked in mL and pints
  • Ergonomically designed for reduced hand fatigue
  • Chemical resistant and multipurpose for various applications
Catalog No. 50-286-6098 Supplier MSC Supplier No. 74254699
The Kwazar Mercury Pro+ Spray Bottle features a robust, ergonomically designed construction for professional applications. It is equipped with a unique dual-action trigger that provides a constant, fine mist with reduced hand fatigue, suitable for various chemical dispensing tasks.
  • Unique double-action trigger provides a constant, fine spray
  • Viton seals resistant to most common chemicals and detergents
  • Level indicator embossed on container marked in mL and pints
  • Ergonomically designed for reduced hand fatigue
  • Chemical resistant and multipurpose for various applications