The Kwazar Spray Bottle features a 1-liter plastic design for mist dispensing. Its unique double-action trigger provides consistent fine spray and reduces hand fatigue. Equipped with Viton® seals, it resists most common chemicals and detergents. An embossed level indicator in milliliters and pints ensures easy measurement. This durable product is ideal for car care, detailing, window tinting, and professional cleaning.
- Double action trigger provides constant fine spray with less hand fatigue.
- Viton® seals resist most common chemicals and detergents.
- Embossed level indicator on container in milliliters and pints.
- Ideal for car care, detailing, window tinting, and professional cleaning.
- Green trigger sprayer, plastic bottle, and nozzle.
- Container has 1 L capacity.
- Mist dispensing type.