Learn More
MSC Kwazar Spray Bottle, Plastic, 1 L, Viton Seals, 1 ea
Catalog No. 502867700
Encompass
This plastic spray bottle from Kwazar features a 1 L capacity and a mist dispensing type. It is equipped with a double-action trigger and Viton seals, making it suitable for various cleaning applications.
  • Features a double action trigger for a constant fine spray with less hand fatigue
  • Equipped with Viton seals resistant to most common chemicals and detergents
  • Includes a level indicator embossed with mL and pint markings
  • Suitable for car care, detailing, window tinting, and professional cleaning applications
  • Constructed from durable plastic
  • Has a 1 L capacity for various liquid solutions
  • Designed for mist dispensing
  • Red trigger sprayer for easy identification
Catalog No. 50-286-7700 Supplier MSC Supplier No. 74255845
This plastic spray bottle from Kwazar features a 1 L capacity and a mist dispensing type. It is equipped with a double-action trigger and Viton seals, making it suitable for various cleaning applications.
  • Features a double action trigger for a constant fine spray with less hand fatigue
  • Equipped with Viton seals resistant to most common chemicals and detergents
  • Includes a level indicator embossed with mL and pint markings
  • Suitable for car care, detailing, window tinting, and professional cleaning applications
  • Constructed from durable plastic
  • Has a 1 L capacity for various liquid solutions
  • Designed for mist dispensing
  • Red trigger sprayer for easy identification