Learn More
MSC Kwazar Spray Bottle with Trigger, 1 L, Plastic, Mist Dispenser, 360° Function
Catalog No. 502837962
Encompass
This spray bottle with a trigger features a 360° function for continuous spraying at any angle, including upside down, ensuring full product utilization. Its double-action trigger provides a constant fine mist, reducing hand fatigue. Viton seals offer resistance to most common chemicals and detergents. A level indicator, embossed with ML and pint markings, is also present.
  • 360° spraying capability for uninterrupted spraying at any angle.
  • Double-action trigger provides consistent fine mist and reduces hand strain.
  • Chemically resistant seals.
  • Level indicator with ML and pint markings.
  • Suitable for car care and detailing.
  • Ideal for cleaning professionals, utilizing the 360° feature.
Catalog No. 50-283-7962 Supplier MSC Supplier No. 74333162
This spray bottle with a trigger features a 360° function for continuous spraying at any angle, including upside down, ensuring full product utilization. Its double-action trigger provides a constant fine mist, reducing hand fatigue. Viton seals offer resistance to most common chemicals and detergents. A level indicator, embossed with ML and pint markings, is also present.
  • 360° spraying capability for uninterrupted spraying at any angle.
  • Double-action trigger provides consistent fine mist and reduces hand strain.
  • Chemically resistant seals.
  • Level indicator with ML and pint markings.
  • Suitable for car care and detailing.
  • Ideal for cleaning professionals, utilizing the 360° feature.