Learn More
Bulbconnection Dot Com MERCURY SHORT ARC HBO100W/2
Catalog No. NC0463378
Encompass
OSRAM 69217 HBO100W/2. Microscopy, Curing/Drying. 100 WATTS; 20 VOLTS; SFA7,5-2 / SFA9-2 BASE TYPE; CLEAR FINISH; 2200 LUMENS; 260 CANDLE POWER; 200 HRS.; 0.394in BULB DIAMETER; 1.693in LIGHT CENTER LEN; 3.543in MAX OVERALL LEN
Catalog No. NC0463378 Supplier Bulbconnection Dot Com Supplier No. 69217
OSRAM 69217 HBO100W/2. Microscopy, Curing/Drying. 100 WATTS; 20 VOLTS; SFA7,5-2 / SFA9-2 BASE TYPE; CLEAR FINISH; 2200 LUMENS; 260 CANDLE POWER; 200 HRS.; 0.394in BULB DIAMETER; 1.693in LIGHT CENTER LEN; 3.543in MAX OVERALL LEN