Learn More
Inorganic Ventures 1µg/mL MERCURY HCl 125mL
SDP
Catalog No. NC1541927
Encompass_Preferred
1ug/mL MERCURY HCl has a matrix of 10% v/v Hydrochloric Acid (CAS# 7647-01-0).
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC1541927 Supplier Inorganic Ventures Supplier No. MSHG1PPM125ML
1ug/mL MERCURY HCl has a matrix of 10% v/v Hydrochloric Acid (CAS# 7647-01-0).
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search