Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mercury standard solution, for AAS, 1 mg/ml Hg in 2 to 5% HNO3

Catalog No. AC196121000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC196121000 100 mL
AC196125000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. AC196121000 Supplier Thermo Scientific Chemicals Supplier No. 196121000
Only null left
Add to Cart
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

CAS: 7783-34-8 | H2HgN2O7 | 342.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mercury standard solution
Packaging Plastic bottle
Name Note 1 mg/ml Hg in 10% HNO3
CAS 7783-34-8, 7697-37-2, 7732-18-5
Density 1.0200g/mL
Specific Gravity 1.02
Physical Form Powder, Granules or Chunks
Molecular Formula H2HgN2O7
Concentration 990 to 1010 μg/mL of Hg(2+) in 2 to 5% HNO3
MDL Number MFCD00149736
Color Black to Gray
Melting Point -3.0°C
Boiling Point 101.0°C
Solubility Soluble in water
InChI Key KVICROHOONHSRH-UHFFFAOYSA-N
SMILES O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name mercury(2+);dinitrate;hydrate
Molecular Weight (g/mol) 342.62
Quantity 100 mL
PubChem CID 3084029
Formula Weight 342.62
Grade AAS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.