Learn More
Cpi International MERCURY STD ELEMENT 100ML
SDP
Catalog No. NC9878594
Encompass
Mercury Standard Element 100mlin 2% HNO3; Contains mercury
Catalog No. NC9878594 Supplier Cpi International Supplier No. S44001000331
Add to Cart
Add to Cart
Mercury Standard Element 100mlin 2% HNO3; Contains mercury