Learn More
Sigma Aldrich Fine Chemicals Biosciences MercuryII iodide ACS reage100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 221090100G
Encompass
Red mercuric oxide possesses the ability to detect high energy radiation (X- and³-rays) and is used in nuclear radiation detectors.
Catalog No. 50-289-7131