Learn More
Sigma Aldrich Fine Chemicals Biosciences MercuryII iodide ACS reage500G
Catalog No. 502903672
Encompass
Red mercuric oxide possesses the ability to detect high energy radiation (X- and³-rays) and is used in nuclear radiation detectors.
Catalog No. 50-290-3672 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 221090500G
Red mercuric oxide possesses the ability to detect high energy radiation (X- and³-rays) and is used in nuclear radiation detectors.