Learn More
Medchemexpress LLC HY-15514A 5mg Medchemexpress, Merestinib dihydrochloride CAS:1206801-37-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15514A5MG
Encompass_Preferred
Medchemexpress, HY-15514A 5mg Merestinib dihydrochloride CAS:1206801-37-7 Merestinib dihydrochloride (LY2801653 dihydrochloride) is a potent, orally bioavailable c-Met inhibitor (Ki=2 nM) with anti-tumor activities. Merestinib dihydrochloride also has potent activity against MST1R (IC50=11 nM), FLT3 (IC50=7 nM), AXL (IC50=2 nM), MERTK (IC50=10 nM), TEK (IC50=63 nM), ROS1, DDR1/2 (IC50=0.1/7 nM) and MKNK1/2 (IC50=7 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1729