Learn More
Selleck Chemical LLC Merestinib dihydrochloride E7462-5MG
SDP
Catalog No. 5000041005
Encompass_Preferred
Merestinib dihydrochloride (LY2801653 dihydrochloride) is a potent orally bioavailable c-Met inhibitor (Ki 2 nM) with anti-tumor activities Merestinib dihydrochloride also has potent activity against MST1R (IC50 11 nM) FLT3 (IC50 7 nM) AXL (IC50 2 nM) MERTK (IC50 10 nM) TEK (IC50 63 nM) ROS1 DDR1/2 (IC50 0 1/7 nM) and MKNK1/2 (IC50 7 nM)
Catalog No. 50-000-41005 Supplier Selleck Chemical LLC Supplier No. E74625MG
Merestinib dihydrochloride (LY2801653 dihydrochloride) is a potent orally bioavailable c-Met inhibitor (Ki 2 nM) with anti-tumor activities Merestinib dihydrochloride also has potent activity against MST1R (IC50 11 nM) FLT3 (IC50 7 nM) AXL (IC50 2 nM) MERTK (IC50 10 nM) TEK (IC50 63 nM) ROS1 DDR1/2 (IC50 0 1/7 nM) and MKNK1/2 (IC50 7 nM)