Learn More
Medchemexpress LLC Merestinib dihydrochloride | 1206801-37-7 | 99.4% | 625.45 g·mol⁻¹ | C30H24Cl2F2N6O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15514A10MG
Encompass_Preferred
Merestinib dihydrochloride is the dihydrochloride salt of merestinib (LY2801653), an orally bioavailable small-molecule multi-kinase inhibitor supplied for preclinical research. It potently inhibits c-Met and has reported activity against several oncogenic kinases, and is provided as a characterized reagent with supporting analytical data.
  • Potent c-Met inhibitor with reported Ki ≈ 2 nM.
  • Activity against multiple kinases including MST1R, FLT3, AXL, MERTK, TEK, ROS1, DDR1/2, and MKNK1/2.
  • Orally bioavailable small-molecule suitable for in vitro and in vivo preclinical studies.
  • Characterized reagent with molecular formula C30H24Cl2F2N6O3 and molecular weight 625.45 g·mol⁻¹.
  • High purity confirmed by LCMS (≈99.4%).
Catalog No. 50-002-86771