Learn More
Selleck Chemical LLC Merestinib (LY2801653) 5mg 1206799-15-6
SDP
Catalog No. 501948141
Encompass_Preferred
Merestinib (LY2801653) is a type-II ATP competitive, slow-off inhibitor of Met (c-Met) tyrosine kinase with a dissociation constant (Ki) of 2 nM, a pharmacodynamic residence time (Koff) of 0.00132 min(-1) and t1/2 of 525 min. Merestinib (LY2801653) also inhibits MST1R, AXL, ROS1, MKNK1/2, FLT3, MERTK, DDR1 and DDR2 with IC50 of 11 nM, 2 nM, 23 nM, 7 nM, 7 nM, 10 nM, 0.1 nM and 7 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8141 Supplier Selleck Chemical LLC Supplier No. S70145MG
Add to Cart
Add to Cart
Merestinib (LY2801653) is a type-II ATP competitive, slow-off inhibitor of Met (c-Met) tyrosine kinase with a dissociation constant (Ki) of 2 nM, a pharmacodynamic residence time (Koff) of 0.00132 min(-1) and t1/2 of 525 min. Merestinib (LY2801653) also inhibits MST1R, AXL, ROS1, MKNK1/2, FLT3, MERTK, DDR1 and DDR2 with IC50 of 11 nM, 2 nM, 23 nM, 7 nM, 7 nM, 10 nM, 0.1 nM and 7 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.