Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian™ Aspergillus fumigatus Immunodiffusion Antigen
GSA_VA

Catalog No. 23030800 Shop All Meridian Bioscience Products
Change view
Click to view available options
Volume (Metric):
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
23-030800 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
When handling blood specimens, adequate measures should be taken to prevent the dissemination of etiologic agents potentially present in the specimen.
Catalog No. 23-030800 Supplier Meridian™ Supplier No. 100401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

When handling blood specimens, adequate measures should be taken to prevent the dissemination of etiologic agents potentially present in the specimen.

Aspergillus fumigatus antigen for use in Fungal Immunodiffusion

  • Reagents is standardized and purified for the in vitro determination of precipitating antibodies to the systemic fungal pathogen Aspergillus fumigatus
  • Reagents is optimized for use in the Ouchterlony diffusion method

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Antigen
For Use With (Application) In-vitro determination of precipitating antibodies to systemic fungal pathogens Aspergillus fumigatus
Storage Requirements 2° to 8°C
Volume (Metric) 1mL
Test Time 24 hr.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.