Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Diagnostics Premier™ Adenoclone
GSA_VA

Catalog No. 14910181 Shop All Meridian Bioscience Products
Change view
Click to view available options
Sample Type:
Cell Culture, Stool
Stool
Sensitivity:
98%
99%
Specificity:
0.98
1
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type Sensitivity Specificity
14-910-181 Cell Culture, Stool 98% 0.98
14-910-180 Stool 99% 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-910-181 Supplier Meridian Bioscience™ Supplier No. 696006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Qualitative ELISA for the detection of human adenoviruses

  • Sensitivity: 98%
  • Specificity: 98%
  • Enzyme immunoassay that utilizes monoclonal antibodies
  • Allows qualitative detection of human adenoviruses direct from stool specimens
  • Results Interpretation: Spectrophotometer/Visual
  • For in vitro use only

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance CE Marked, FDA
Content And Storage 2°C to 8°C
CPT Code 87301
Sample Type Cell Culture, Stool
Sensitivity 98%
Type Adenovirus Test Kit
For Use With (Application) Detection of enteric adenovirus serotypes 40 and 41 in Human fecal specimens
Includes 48 Microwells, 50mL sample diluent/negative control, 60mL Enzyme conjugate, 2mL positive control, 6mL substrates and stop solution, microwell holder, 48 sample transfer pipettes
Quantity 48 Tests
Specificity 0.98
Test Time 75 min.
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.