Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ ImmunoCard STAT!™ EHEC
GSA_VA

Catalog No. 23043200 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-043-200 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-200 Supplier Meridian Bioscience™ Supplier No. 751630
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

30 Test devices packaged in individual foil pouches with desiccants; sample diluent (negative control) in a plastic dropper; positive control in a plasic dropper vial; and 150μL disposable plastic transfer pipets

Rapid lateral-flow test that detects and differentiates both E. coli Shiga toxin 1 and Shiga toxin 2

  • CLIA Complexity: Non-waived (not verified)
  • CPT Code: 87899
  • Sensitivity: 93.8%
  • Specificity: 99.7%
  • Test results in 20 minutes
  • Reports two results, and differentiates between Shiga toxin 1 and Shiga toxin 2
  • Highly sensitive detection of toxins contributes to prevention of serious illnesses such as HUS

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Clia Complexity Non-waived
Detectable Analytes E. coli Shiga Toxin 1 and 2
Type ImmunoCard Stat™ EHEC Test Kit
Includes 30 Test devices, sample diluent (negative control) in a Plastic dropper, positive control in a Plasic dropper vial and 150μL disposable Plastic transfer pipets
Quantity 30 Tests
Test Time 20 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.