Learn More
Meridian Bioscience™ OSOM hCG Combo Test, 25 Tests
Catalog No. 23030150 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
25 Tests
Catalog No. Quantity
23-030-150 25 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-030-150 Supplier Meridian Bioscience™ Supplier No. 124
Add to Cart
Add to Cart
This item is not returnable. View return policy
A simple, rapid immunoassay for the qualitative detection of human chorionic gonadotropin (hCG) in urine or serum as an aid in the early determination of pregnancy
  • CLIA Complexity: Urine: Waived; Serum: Non-Waived
  • Sensitivity: 10mIU/mL serum, 20mIU/mL urine
  • Simple, easy-to-read results
  • Rapid results: three minutes for urine; five minutes for serum
Certifications
CE marked
Specifications
Certifications/Compliance CE Marked
CPT Code 81025, 84703
Clia Complexity Waived, Non-waived
Detectable Analytes hCG
Sample Type Serum, Urine
Type hCG Combo Test
Quantity 25 Tests
Test Time 3 min. for urine, 5 min. for serum
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov