Learn More
Meridian Bioscience™ ImmunoCard™ STAT! Mono Test Kit
GSA_VA
Catalog No. 23030154 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
25 Tests
Catalog No. Quantity
23-030-154 25 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-030-154 Supplier Meridian Bioscience™ Supplier No. 755725
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Includes
Test cards, positive control, negative control, developer buffer and transfer pipets
Unitary one-step immuno-chromatographic assay for the qualitative detection of infectious mononucleosis heterophile antibodies
  • Human serum, plasma or whole blood
  • Easy and unitary procedure
  • Results within 10 minutes
  • Positive result: one pink-red colored band in the test and control zones
  • Negative result: one pink-red colored band in control zone only
  • Built-in procedural control on each card
  • Ready-to-use reagents that can be stored at room temperature
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Detectable Analytes Mononucleosis Heterophil Antibodies
Format Kit
Type Mononucleosis Test Kit
Includes Test cards, positive control, negative control, developer Buffer and transfer pipets
Quantity 25 Tests