Learn More
Meridian Bioscience™ ImmunoCard™ STAT! Strep A Test Kit
GSA_VA
Catalog No. 23030153 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
50 Tests
Catalog No. Quantity
23-030-153 50 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-030-153 Supplier Meridian Bioscience™ Supplier No. 755250
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Test packs (one test strip and one tablet in the extraction tube), extraction reagent, throat swabs, positive and negative controls
Lateral flow one-step immunoassay for the rapid qualitative detection of Group A Streptococcal antigen from human throat swabs
  • Rapid detection: results within 10 minutes
  • Easy unitary procedure
  • Internal control of the procedure on each test strip
  • Ready-to-use reagents
  • Use only dacron or rayon tipped sterile swabs with plastic shafts; swabs can be stored at room temperature up to four hours or refrigerated up to 24 hours
  • Requires 9 to 11 drops of extraction reagent placed into the extraction tube
  • Positive result: one band in the control zone and one band in the test zone; negative result: one band in the control zone only
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Certifications/Compliance CE Marked
CPT Code 87880
Detectable Analytes Streptococci (Group A)
Sample Type Culture, Throat Swab
Type Streptococcus A Test Kit
Includes Test packs (one test strip and one tablet in the extraction tube), extraction reagent, throat swabs, positive and negative control
Quantity 50 Tests
Test Time 10 min.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov