Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ MeriFluor™ Cryptosporidium/Giardia Test Kit Accessories: 3-Well Treated Slides
GSA_VA

Catalog No. 23315120 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
25 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-315-120 25 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-315-120 Supplier Meridian Bioscience™ Supplier No. R2206
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Come in a pack of 25

  • For use with MeriFluor Cryptosporidium/Giardia Test Kit

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Format Treated Slide
Type 3-wells Treated Slide
Quantity 25 Pack
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.