Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ MeriFluor™ Cryptosporidium/Giardia Test Kit
GSA_VA

Catalog No. 14910115 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-115 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-910-115 Supplier Meridian Bioscience™ Supplier No. 250050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Detection reagent, counterstain, wash buffer, mounting medium, treated slides, transfer loops, positive and negative controls for 50 tests

One-step procedure

  • CLIA Complexity: non-waived
  • CPT Codes: 87269, 87272
  • Immunofluorescent assay
  • Identifies the presence of both Cryptosporidium oocysts and Giardia cysts in stool specimens
  • Test results within 30 minutes
  • Uses monoclonal antibodies to ensure increased sensitivity of 100%
  • Enhanced differentiation due to contrast of the apple-green fluorescence of the parasite against red-orange background makes rare organisms stand out
  • For in vitro diagnostic use only

Order Info

Additional 3-well treated slides available separately (Cat. No. 23-315-120).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 8°C
CPT Code 87269, 87272
Clia Complexity Non-waived
Detectable Analytes Cryptosporidium Oocyst, Giardia Cyst
Sensitivity 1
Type Cryptosporidium/Giardia Test Kit
Includes Detection reagent, counterstain, Wash Buffer, mounting medium, treated slides, transfer loops, positive and negative control
Quantity 50 Tests
Specificity 1
Test Time 30 min.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.