Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Para-Pak™ CON-Trate™ Concentration Systems
GSA_VA

Catalog No. 14910165 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-165 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-910-165 Supplier Meridian Bioscience™ Supplier No. 960200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Caps, filtration devices, 15mL graduated centrifuge tubes, CON-Trate Reagent A (MucoPenX) and Reagent B (ethyl acetate)

Injury-free parasite retrieval

  • Designed to optimize parasitic concentration/recovery and minimize specimen preparation time
  • Kits of 50 or 500 tests
  • Filtration devices also available separately

Order Info

Packaged in a workstation design to reduce procedure time

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Stool Concentration System
Certifications/Compliance CE
Includes Cap, filtration devices, 15mL graduated centrifuge tubes, Con-Trate™ reagent A (MucoPenX) and reagent B (Ethyl Acetate)
Packaging Workstation
Quantity 200 Tests
Storage Requirements 20°C to 30°C
For Use With (Application) Preserved and unpreserved stool
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.