Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Para-Pak™ Macro-CON™ Accessories

Catalog No. 149101149 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
1 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-1149 1 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-910-1149 Supplier Meridian Bioscience™ Supplier No. 970001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use in Macro-CON concentration of eggs, larvae, and cysts

Specifications

Detectable Analytes Cysts, Eggs, Larvae
Type Macro-CON™ Surfactant
For Use With (Application) Macro-CON™ concentration of eggs, larvae and cysts
Quantity 1 Pack
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.